Merck
Treatment of phosphogypsum waste produced from phosphate ore processing.
Journal of hazardous materials (2012-12-01)
H El-Didamony, H S Gado, N S Awwad, M M Fawzy, M F Attallah
RESUMEN
Phosphogypsum (PG), primary byproduct from phosphoric acid production, is accumulated in large stockpiles and occupies vast areas of land. Phosphogypsum is a technologically enhanced naturally occurring radioactive material (TE-NORM) that contains radionuclides from (238)U and (232)Th decay series which are of most radio-toxicity. The reduction in concentration of radionuclides content from PG was based on leaching of (226)Ra, (210)Pb, (238)U and (40)K using tri-butyl phosphate (TBP) and tri-octyl phosphine oxide (TOPO) in kerosene. The factors which affect the leaching process such as contact time, concentration of the solvent and temperature were optimized. Based on the experimental results, about 92.1, 88.9, 83.4, 94.6% of (226)Ra, (210)Pb, (238)U and (40)K respectively were successfully removed from the PG. The reduction in the concentration of radionuclides was accompanied by reduction in the concentration of rare earth elements (∑REE) equals to 80.1%. Using the desired organic extractant under optimum conditions for treatment of the PG waste leads to obtain a decontaminated product that can be safely used in many industrial applications.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Drierite, with indicator, 8 mesh
Sigma-Aldrich
Drierite, with indicator, 10-20 mesh
Sigma-Aldrich
Calcium sulfate dihydrate, ReagentPlus®, ≥99%
Sigma-Aldrich
Calcium sulfate, −325 mesh, 99%
Sigma-Aldrich
Trioctylphosphine oxide, ReagentPlus®, 99%
Sigma-Aldrich
Calcium sulfate hemihydrate, purum, ≥97.0%
Sigma-Aldrich
Calcium sulfate dihydrate, ACS reagent, 98%
Sigma-Aldrich
Drierite, without indicator, 8 mesh
Sigma-Aldrich
Drierite, with indicator, 4 mesh
Sigma-Aldrich
Fosfato de tributilo, ≥99%
Sigma-Aldrich
Fosfato de tributilo, 97%
Sigma-Aldrich
Drierite, with indicator, 6 mesh
Sigma-Aldrich
Trioctylphosphine oxide, technical grade, 90%
Sigma-Aldrich
Calcium sulfate dihydrate, puriss., meets analytical specification of NF, E 516, 99.0-101.0% (based on anhydrous substance)
Sigma-Aldrich
Calcium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Drierite, without indicator, 4 mesh
Sigma-Aldrich
Fosfato de tributilo, puriss., ≥99.0% (GC)
Supelco
Fosfato de tributilo, Pharmaceutical Secondary Standard; Certified Reference Material
Fosfato de tributilo, European Pharmacopoeia (EP) Reference Standard
Supelco
Fosfato de tributilo, for extraction analysis, ≥99.0% (GC)
Sigma-Aldrich
Trioctylphosphine oxide, for extraction analysis, ≥98.5%