Merck
  • Solubilization of parabens in aqueous Pluronic solutions: investigating the micellar growth and interaction as a function of paraben composition.
Solubilization of parabens in aqueous Pluronic solutions: investigating the micellar growth and interaction as a function of paraben composition.
The journal of physical chemistry. B (2012-12-06)
M Khimani, R Ganguly, V K Aswal, S Nath, P Bahadur
RESUMEN
The influence of methyl paraben (MP) and butyl paraben (BP) on the aggregation characteristics of Pluronics in an aqueous medium has been investigated by DLS, SANS, viscometry, and fluorescence measurement techniques. Parabens are extensively used as preservatives in cosmetic, pharmaceutical, and food products. In this paper, we show that their influence on the restructuring and growth of Pluronics micelles vary quite significantly with their aqueous solubility and with the composition of Pluronics. In the case of P105 and P104, MP reduces the sphere-to-rod transition temperature down to room temperature, but BP with significantly less aqueous solubility than MP suppresses such micellar transition and leads to the formation of micellar clusters due to the onset of intermicellar attractive interaction. In the case of more hydrophobic Pluronic P103, on the other hand, both MP and BP are able to induce rapid room temperature sphere-to-rod micellar growth, which is not observed in the presence of water structure making salts like NaCl and Na(3)PO(4). These observations have been attributed to modulation of growth and restructuring processes of the Pluronic micelles arising due to different locations of parabens within the micellar corona as determined by their aqueous solubility and the hydrophobicity of the Pluronics.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Metil 4-hidroxibenzoato, Pharmaceutical Secondary Standard; Certified Reference Material
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Metil 4-hidroxibenzoato, ReagentPlus®, ≥99.0%, crystalline
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99.0% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Methyl Paraben, PESTANAL®, analytical standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Butylparaben, Pharmaceutical Secondary Standard; Certified Reference Material
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Metil 4-hidroxibenzoato, BioReagent, suitable for insect cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Metil 4-hidroxibenzoato, ≥99%, FCC
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Metil 4-hidroxibenzoato, European Pharmacopoeia (EP) Reference Standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Butyl 4-hydroxybenzoate, ≥99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Metil 4-hidroxibenzoato, BioXtra, ≥99.0% (titration)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Metil 4-hidroxibenzoato, analytical standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Metil 4-hidroxibenzoato, tested according to Ph. Eur.
En este momento no podemos mostrarle ni los precios ni la disponibilidad