Merck
Quantitative investigation of scintillator intensification for light and electron microscope radioautography.
Histochemistry (1980-01-01)
B M Kopriwa
RESUMEN
Several reports have indicated that scintillators enhance the intensity of radioautographic reactions. If this were the case, the use of scintillators would shorten the exposure time of radioautographs and the production of radioautographs would be accelerated. It was, therefore, decided to examine the effect of scintillators on the intensity of radioautographs in a quantitative manner over uniformly labeled test specimens. Reaction intensities with or without scintillators were compared using Kodak NTB2 emulsion for light microscopy and Ilford L4 and Sakura NR-H2 emulsion for electron microscopy. In addition, the background fog was quantitatively evaluated. No increase in the intensity of the radioautographs was observed under the following conditions: pretreatment of tissue sections with scintillator solutions before emulsion coating and exposure; treatment with scintillator solutions after emulsion coating but before exposure; treatment with scintillator solutions during exposure; contact with a plastic scintillator film throughout exposure. Heightened reaction intensity was obtained only when scintillators were applied during the histological processing of tissues. The impregnation of tissue blocks with PPO (2,5-Diphenyloxazole) and POPOP (1,4-bis-2-(5-phenyloxazolyl)-benzene) during dehydration, infiltration and Epon embedding produced a 20% increase of reaction intensity in light microscope radioautographs using NTB2 emulsion and a 75% increase in electron microscope radioautographs using L4 emulsion.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
1,4-Bis(5-phenyl-2-oxazolyl)benzene, BioReagent, suitable for scintillation
Sigma-Aldrich
1,4-Bis(5-phenyl-2-oxazolyl)benzene, suitable for scintillation, 98%