Merck
Active transport of nitrofurantoin across the mammary epithelium in vivo.
The Journal of pharmacology and experimental therapeutics (1997-02-01)
F W Kari, R Weaver, M C Neville
RESUMEN
Nitrofurantoin is a commonly used urinary tract antibiotic that has been found at high concentrations in human milk. In vivo studies in rats were carried out to determine the mechanism by which this drug crosses the mammary epithelium. Lactating rats were gavage-fed with nitrofurantoin, and their milk and plasma levels of the antibiotic were measured at intervals up to 8 hr. The average milk-to-plasma (M/P) ratio, calculated from the areas under the milk and plasma curves, respectively, was 23 compared with a ratio predicted to be about 0.3 on the basis of lipid partitioning and protein binding determinations. M/P ratios for two nitrofurantoin congeners were also calculated. The neutral compound furazolidone had a M/P ratio of about 1, as predicted, whereas the basic compound furaltadone had a M/P ratio of 3.49 compared with a predicted ratio of 1.4. These data suggest that nitrofurantoin and, to a lesser extent, furaltadone are actively transported across the mammary epithelium into milk.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Furaltadone
Supelco
Furaltadone, VETRANAL®, analytical standard