Merck
Metabolism of dl-praeruptorin A in rat liver microsomes using HPLC-electrospray ionization tandem mass spectrometry.
Archives of pharmacal research (2011-09-13)
Hang Ruan, Zhen Zhang, Xin-fang Liang, Yan Fu, Mei-qin Su, Qi-lin Liu, Xiu-min Wang, Xuan Zhu
RESUMEN
dl-Praeruptorin A (Pd-Ia) is the major active constituent of the traditional Chinese medicine Peucedanum praeruptorum Dunn. Recently it has been identified as a novel agent in the treatment and prevention of cardiovascular diseases. Accordingly, we investigated the metabolism of Pd-Ia in rat liver microsomes. The involvement of cytochrome P450 (CYP) and CYP isoforms were identified using a CYP-specific inhibitor (SKF-525A), CYP-selective inhibitors (α-naphthoflavone, metyrapone, fluvastatin, quinidine, disulfiram, ketoconazole and ticlopidine) and CYP-selective inducers (phenobarbital, dexamethasone and β-naphthoflavone). Residual concentrations of the substrate and metabolites were determined by HPLC, and further identified by their mass spectra and chromatographic behavior. These experiments showed that CYP450 is involved in Pd-Ia metabolism, and that the major CYP isoform responsible is CYP3A1/2, which acts in a concentration-dependent manner. Four Pd-Ia metabolites (M1, M2, M3, and M4) were detected after incubation with rat liver microsomes. Hydroxylation was the primary metabolic pathway of Pd-Ia, and possible chemical structures of the metabolites were identified. Further research is now needed to link the metabolism of Pd-Ia to its drug-drug interactions.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Proadifen hydrochloride, analytical standard, ≥95%