Merck
Dual sites of protein initiation control the localization and myristoylation of methionine sulfoxide reductase A.
The Journal of biological chemistry (2010-04-07)
Geumsoo Kim, Nelson B Cole, Jung Chae Lim, Hang Zhao, Rodney L Levine
RESUMEN
Methionine sulfoxide reductase A is an essential enzyme in the antioxidant system, which scavenges reactive oxygen species through cyclic oxidation and reduction of methionine and methionine sulfoxide. In mammals, one gene encodes two forms of the reductase, one targeted to the cytosol and the other to mitochondria. The cytosolic form displays faster mobility than the mitochondrial form, suggesting a lower molecular weight for the former. The apparent size difference and targeting to two cellular compartments had been proposed to result from differential splicing of mRNA. We now show that differential targeting is effected by use of two initiation sites, one of which includes a mitochondrial targeting sequence, whereas the other does not. We also demonstrate that the mass of the cytosolic form is not less than that of the mitochondrial form; the faster mobility of cytosolic form is due to its myristoylation. Lipidation of methionine sulfoxide reductase A occurs in the mouse, in transfected tissue culture cells, and even in a cell-free protein synthesis system. The physiologic role of myristoylation of MsrA remains to be elucidated.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Protease Inhibitor Cocktail powder, for general use, lyophilized powder
Sigma-Aldrich
Mixed Bed Resin hydrogen and hydroxide form, DNA grade, BioReagent, Molecular Biology