Merck
Acute effects of carbofuran in workers of two pesticide plants.
La Medicina del lavoro (1999-04-28)
J Huang, S Zhang, M Ding, A Zhou, J Zhang, J Zhang
RESUMEN
This paper investigates the acute effects of carbofuran in workers of two pesticide-formulating plants. Mean airborne carbofuran concentrations ranged from 0.025 to 1.115 mg/m3 in plant A and from 0.018 to 0.067 mg/m3 in plant B, respectively. In workers of plant A the post-shift blood cholinesterase activity was significantly reduced, compared to pre-shift values. No difference in blood cholinesterase activity was found between pre- and post-shift values in workers of plant B. During the investigation, 25 cases of acute carbofuran poisoning were diagnosed by their clinical picture and depressed cholinesterase activity in blood. Usual symptoms included dizziness, weakness, blurred vision, nausea and sweating. Pallor, epigastric pain, vomiting and chest tightness occurred only in a few cases. Myosis was recorded in 24 cases. Fasciculation of muscle gastrocnemius induced by percussion was found in 6 cases, and four of them had also fasciculation of muscle orbicularis oculi. Inhibition of cholinesterase activity in the blood was related with the clinical features; however, the inhibition was rapidly reversible. In most cases, recovery was complete within 2-3 hours, with or without atropine treatment, after the subjects were removed from exposure. Rapid onset, mild illness and quick recovery are typical characteristics of occupational acute carbofuran poisoning.
MATERIALES
Número de producto
Marca
Descripción del producto
Supelco
Carbofuran-3-hydroxy, PESTANAL®, analytical standard