Merck
Anti-inflammatory activity of c(ILDV-NH(CH2)5CO), a novel, selective, cyclic peptide inhibitor of VLA-4-mediated cell adhesion.
British journal of pharmacology (1999-06-18)
D Haworth, A Rees, P J Alcock, L J Wood, A S Dutta, J J Gormley, H B Jones, A Jamieson, C F Reilly
RESUMEN
1. Small, N- to C-terminal cyclized peptides containing the leucyl-aspartyl-valine (LDV) motif from fibronectin connecting segment-1 (CS-1) have been investigated for their effects on the adhesion of human T-lymphoblastic leukaemia cells (MOLT-4) to human plasma fibronectin in vitro mediated by the integrin Very Late Antigen (VLA)-4 (alpha4beta1, CD49d/CD29). 2. Cyclo(-isoleucyl-leucyl-aspartyl-valyl-aminohexanoyl-) (c(ILDV-NH(CH2)5CO)) was approximately 5 fold more potent (IC50 3.6+/-0.44 microM) than the 25-amino acid linear CS-1 peptide. Cyclic peptides containing two more or one less methylene groups had similar potency to c(ILDV-NH(CH2)5CO) while a compound containing three less methylene groups, c(ILDV-NH(CH2)2CO), was inactive at 100 microM. 3. c(ILDV-NH(CH2)5CO) had little effect on cell adhesion mediated by two other integrins, VLA-5 (alpha5,beta1, CD49e/CD29) (K562 cell adhesion to fibronectin) or Leukocyte Function Associated molecule-1 (LFA-1, alphabeta2, CD11a/CD18) (U937 cell adhesion to Chinese hamster ovary cells transfected with intercellular adhesion molecule-1) at concentrations up to 300 microM. 4. c(ILDV-NH(CH2)5CO) inhibited ovalbumin delayed-type hypersensitivity or oxazolone contact hypersensitivity in Balb/c mice when dosed continuously from subcutaneous osmotic mini-pumps (0.1-10 mg kg(-1) day(-1)). Maximum inhibition (approximately 40%) was similar to that caused by the monoclonal antibody PS/2 (7.5 mg kg(-1) i.v.) directed against the alpha4 integrin subunit. 5. c(ILDV-NH(CH2)5CO) also inhibited oxazolone contact hypersensitivity when dosed intravenously 20 h after oxazolone challenge (1-10 mg kg(-1)). Ear swelling was reduced at 3 h and 4 h but not at 1 h and 2 h post-dose (10 mg kg(-1)). 6. Small molecule VLA-4 inhibitors derived from c(ILDV-NH(CH2)5CO) may be useful as anti-inflammatory agents.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Fibronectin Type III Connecting Segment Fragment 1-25, ≥90% (HPLC)