Merck
Biomonitoring at the UK Health and Safety Laboratory.
International journal of hygiene and environmental health (2007-02-27)
J Cocker, K Jones, J Morton, H J Mason
RESUMEN
The UK Health and Safety Laboratory (HSL) provides research and analytical support to the Health and Safety Executive, other Government Departments and employers. In the area of biomonitoring HSL conducts research studies and provides an analytical service for regular surveillance of worker exposure to hazardous substances. This paper gives brief examples of how data from such studies can be used to develop biological monitoring guidance values for isocyanates, polycyclic aromatic hydrocarbons and hexavalent chromium. In addition, a study of occupational exposure to copper chrome arsenic wood preservatives is briefly described to show how biological monitoring can be used for post-approval surveillance of a biocide.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
4,4′-Methylene-bis(2-chloroaniline), 85%