Merck
  • Lack of NMDA-AMPA interaction in antidepressant-like effect of CGP 37849, an antagonist of NMDA receptor, in the forced swim test.
Lack of NMDA-AMPA interaction in antidepressant-like effect of CGP 37849, an antagonist of NMDA receptor, in the forced swim test.
Journal of neural transmission (Vienna, Austria : 1996) (2008-10-08)
Małgorzata Dybała, Agata Siwek, Ewa Poleszak, Andrzej Pilc, Gabriel Nowak
RESUMEN
The NMDA receptor antagonist, CGP 37849-induced reduction in immobility time in the forced swim test in mice was not antagonized by pre-treatment with the AMPA receptor antagonist NBQX. This is the first demonstration of the antidepressant effect of the NMDA antagonist not being dependent on the AMPA transmission.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
CGP 37849, ≥98% (HPLC)