Merck
(S)-alpha-methyl,alpha-amino acids: a new stereocontrolled synthesis.
Amino acids (2009-04-14)
Daniele Balducci, Ilaria Lazzari, Magda Monari, Fabio Piccinelli, Gianni Porzi
RESUMEN
A new and convenient stereocontrolled synthesis of the optically pure (S)-alpha-methyl,alpha-amino acids 6(a-d) that exploits the chiral synthon 1,4-N,N-[(S)-1-phenylethyl]-piperazine-2,5-dione (1) is described. The (S)-1-phenylethyl group, bonded to each of the N-atoms of the 2,5-diketopiperazine, acts as a chiral inductor in the first alkylation, while the steric hindrance appears to be the determining factor of stereocontrol in third and forth alkylation.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
DL-Homophenylalanine, 98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
L-Homophenylalanine hydrochloride, 97%
En este momento no podemos mostrarle ni los precios ni la disponibilidad