Merck
Gene cloning and catalysis features of a new mannitol-1-phosphate dehydrogenase (BbMPD) from Beauveria bassiana.
Carbohydrate research (2009-11-10)
Zheng-Liang Wang, Sheng-Hua Ying, Ming-Guang Feng
RESUMEN
A long-chain mannitol-1-phosphate dehydrogenase (MPD) was characterized for the first time from fungal entomopathogen Beauveria bassiana by gene cloning, heterogeneous expression and activity analysis. The cloned gene BbMPD consisted of a 1334-bp open reading frame (ORF) with a 158-bp intron and the 935-bp upstream and 780-bp downstream regions. The ORF-encoded 391-aa protein (42kDa) showed less than 75% sequence identity to 17 fungal MPDs documented and shared two conserved domains with the fungal MPD family at the N- and C-terminus, respectively. The new enzyme was expressed well in the Luria-Bertani culture of engineered Escherichia coli BL21 by 16-h induction of 0.5 mM isopropyl 1-thio-beta-d-galactopyranoside at 20 degrees C after 5-h growth at 37 degrees C. The purified BbMPD exhibited a high catalytic efficiency (k(cat)/K(m)) of 1.31 x 10(4) mM(-1)s(-1) in the reduction of the highly specific substrate d-fructose-6-phosphate to d-mannitol-1-phosphate. Its activity was maximal at the reaction regime of 37 degrees C and pH 7.0 and was much more sensitive to Cu(2+) and Zn(2+) than to Li(+) and Mn(2+). The results indicate a crucial role of BbMPD in the mannitol biosynthesis of B. bassiana.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
D-Mannitol 1-phosphate lithium salt, ≥95% (TLC)