Merck
Wet air oxidation of epoxy acrylate monomer industrial wastewater.
Journal of hazardous materials (2010-03-09)
Shaoxia Yang, Zhengqian Liu, Xiaohui Huang, Beiping Zhang
RESUMEN
Epoxy acrylate monomer industrial wastewater contained highly concentrated and toxic organic compounds. The wet air oxidation (WAO) and catalytic wet air oxidation (CWAO) were used to eliminate pollutants in order to examine the feasibility of the WAO/CWAO as a pre-treatment method for the industrial wastewater. The results showed that in the WAO 63% chemical oxygen demand (COD) and 41% total organic carbon (TOC) removals were achieved and biological oxygen demand (BOD(5))/COD ratio increased from 0.13 to 0.72 after 3h reaction at 250 degrees C, 3.5MPa and the initial concentration of 100g(COD)/L. Among homogenous catalysts (Cu(2+), Fe(2+), Fe(3+) and Mn(2+) salts), Cu(2+) salt exhibited better performance. CuO catalyst was used in the CWAO of the wastewater, COD and TOC conversion were 77 and 54%, and good biodegradability was achieved. The results proved that the CWAO was an effective pre-treatment method for the epoxy acrylate monomer industrial wastewater.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Bisphenol A glycerolate (1 glycerol/phenol) diacrylate, contains MEHQ as inhibitor