Merck
TiC nanoparticles-chitosan composite film for the direct electron transfer of myoglobin and its application in biosensing.
Bioelectrochemistry (Amsterdam, Netherlands) (2012-02-22)
Minzhi Wang, Qinglin Sheng, Dawei Zhang, Yaping He, Jianbin Zheng
RESUMEN
We report on the direct electrochemistry of myoglobin (Mb) immobilized on a composite matrix based on chitosan (CHIT) and titanium carbide nanoparticles (TiC NPs) underlying on glassy carbon electrode (GCE). The cyclic voltammetry and electrochemical impedance spectroscopy were used to characterize the modified electrode. In deaerated buffer solutions, the cyclic voltammetry of the composite films of Mb-TiC NPs-CHIT showed a pair of well-behaved redox peaks that are assigned to the redox reaction of Mb, confirming the effective immobilization of Mb on the composite film. The electron transfer rate constant was estimated to be 3.8 (±0.2)·s(-1), suggested that the interaction between the protein and certain electrode surfaces may mimic some physiological situations and may elucidate the relationship between the protein structures and biological functions. The linear dynamic range for the detection of hydrogen peroxide was 0.5-50 μM with a correlation coefficient of 0.999 and the detection limit was estimated at about 0.2 μM (S/N=3). The calculated apparent Michaelis-Menten constant was 0.07 (±0.01) mM, which suggested a high affinity of the redox protein-substrate. The immobilized Mb in the TiC NPs-CHIT composite film retained its bioactivity. Furthermore, the method presented here can be easily extended to immobilize and obtain the direct electrochemistry of other redox enzymes or proteins.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Titanium(IV) carbide, powder, <4 μm, ≥99% (Ti)
Sigma-Aldrich
Titanium carbide, nanopowder, ≤200 nm particle size (SEM)
Sigma-Aldrich
Titanium(IV) carbide, −325 mesh, 98%