Merck
[Phenolic acid derivatives from Alchornea trewioides].
Yao xue xue bao = Acta pharmaceutica Sinica (2012-09-21)
Ri-Dong Qin, Wei Cheng, Qing-Ying Zhang, Hong Liang
RESUMEN
To study the chemical constituents of Alchornea trewioides, silica gel column chromatography, Sephadex LH-20, reverse phase ODS column chromatography, MCI and semi-preparative HPLC were used to separate the 95% EtOH extract of the root of Alchornea trewioides. The structures were elucidated on the basis of spectroscopic studies including ESI-TOF-MS, 1H NMR, 13C NMR, HSQC and HMBC. Eight phenolic acids were obtained and identified as 1-O-galloyl-6-O-vanilloyl-beta-glucose (1), gallic acid (2), ethyl gallate (3), syringic acid (4), glucosyringic acid (5), erigeside C (6), 3, 4-dimethoxyphenyl-(6'-O-alpha-L-rhamnosyl)-beta-D-glucopyranoside (7) and 3, 4, 5-trimethoxyphenyl-(6'-O-galloyl)-O-beta-D-glucopyranoside (8). Among them, compound 1 is a new compound, compounds 4-8 are isolated from the genus Alchornea for the first time, and the others are isolated from the plant for the first time.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ethyl gallate, antioxidant, ≥96.0% (HPLC)