Merck
  • Contribution of serotonin type 3 receptors in the successful extinction of cued or contextual fear conditioned responses: interactions with GABAergic signaling.
Contribution of serotonin type 3 receptors in the successful extinction of cued or contextual fear conditioned responses: interactions with GABAergic signaling.
Reviews in the neurosciences (2012-10-23)
Su Mi Park, Cedric L Williams
RESUMEN
The changes in serotonin type 3 (5-HT) receptor activity influence memory and emotional regulation,the two essential components underlying the successful extinction of conditioned fear. These studies determined if blocking 5-HT3 receptors with granisetron influences the extinction of fear in male Sprague-Dawley rats. In Experiment 1, preextinction granisetron (0.5 or 1.0 mg/kg intraperitoneally) d~d not affect cued extinction learning but enhanced memory for the extinction session on retention test given 24 and 48 h later. In Experiment 2, granisetron injections given on days 1 and 4 during 6 days of extinction training reduced fear produced by contextual fear conditioning. Experiments 3 and 4 examined if 5-HT3 antagonists influence extinction memory by interactions with y-aminobutyric acid (GABA). The expression of the GABA receptor clustering protein gephyrin was significantly A elevated in the amygdala after cued fear extinction training and a subsequent extinction retention test given 24 h later. Gephyrin expression in the hippocampus but not in the amygdala or the medial prefrontal cortex was significantly reduced after contextual fear extinction sessions given 1 or 5 days after training. The current studies reveal the beneficial effects of 5-HT3 receptor activity in improving new learning associated with extinction of fearful memories and suggest that these actions could be mediated through influences on central GABAergic systems.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Granisetron hydrochloride, ≥98% (HPLC), solid
Granisetron hydrochloride, European Pharmacopoeia (EP) Reference Standard