Merck
[Excretion of 5-hydroxyindole-3-acetic and 5-methoxyindole-3-acetic acids in cancer patients].
Voprosy onkologii (1987-01-01)
T I Sergeeva, T I Raushenbakh, M O Shevchenko, V G Rybal'chenko
RESUMEN
Daily urinary excretion of 5-hydroxy-3-indoleacetic acid (5-HIAA) and 5-methoxyindole-3-acetic acid (5-MIAA) was studied by chromatography-mass-spectrometry in patients with cancer and healthy subjects. A considerable increase in urinary 5-MIAA-excretion was shown in patients with cancer of the stomach, rectum and lung. In some cases, a correlation was found between high urine levels of carcinogenic 5-MIAA and similarly high levels of 5-HIAA excretion with urine.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
5-Methoxy-3-indoleacetic acid, 98%