Merck
Travellers' diarrhoea: prevention by chemoprophylaxis.
Scandinavian journal of gastroenterology. Supplement (1983-01-01)
A C Turner
RESUMEN
The chemoprophylaxis of travellers' diarrhoea by iodochlorhydroxyquinoline combinations, neomycin and phthalsulphathiazole, furazolidone, bismuth salicylates and streptotriad is reviewed. Streptotriad is regarded as a highly effective prophylactic against travellers' diarrhoea. Some elementary food hygiene precautions are detailed.