Merck
Inhibition of human CD4(+)CD25(+high) regulatory T cell function.
Journal of immunology (Baltimore, Md. : 1950) (2002-11-22)
Clare Baecher-Allan, Vissia Viglietta, David A Hafler
RESUMEN
CD4(+)CD25(+high) T cells are potent regulators of autoreactive T cells. However, it is unclear how regulatory CD4(+)CD25(+high) cells discriminate between desirable inflammatory immune responses to microbial Ags and potentially pathologic responses by autoreactive T cells. In this study, an in vitro model was created that allowed differential activation of regulatory CD4(+)CD25(+high) and responder CD4(+) T cells. If CD4(+)CD25(+high) regulatory cells were strongly activated, they maintained suppressive effector function for only 15 h, while stimulation with weaker TCR stimuli produced regulatory cells that were suppressive until 60 h after activation. In contrast, strongly activated CD4(+) responder T cells were resistant to regulation at all time points, while weakly stimulated CD4(+) cells were sensitive to suppression until 38 or 60 h after activation depending upon the strength of the stimulus. The extent of suppression mediated by CD4(+)CD25(+high) cells also depended on the strength of stimulation in an Ag-specific system. Thus, the stronger the TCR signal, the more rapidly and more completely the responder cells become refractory to suppression.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Avidin–Peroxidase, buffered aqueous solution