- Separation of beta-carotene and lycopene geometrical isomers in biological samples.
Separation of beta-carotene and lycopene geometrical isomers in biological samples.
Clinical chemistry (1993-05-01)
W Stahl, A R Sundquist, M Hanusch, W Schwarz, H Sies
PMID8485871
RESUMEN
The analysis of beta-carotene and lycopene, the two predominant carotenoids in human serum and tissues, was extended to the level of geometrical (cis-trans) isomers by using an improved reversed-phase HPLC methodology. We separated five geometrical isomers of beta-carotene and seven of lycopene in human serum and tissues. 13-cis-beta-Carotene was identified as the predominant cisisomer in human serum, contributing about 5% to total beta-carotene. In tissue, however, considerable amounts of 9-cis- and traces of 15-cis-beta-carotene were also detected. In contrast to beta-carotene, the lycopene isomer patterns in human serum and tissues are quite similar.
MATERIALES
Número de producto
Marca
Descripción del producto
En este momento no podemos mostrarle ni los precios ni la disponibilidad