Merck
N-acyl homoserine lactone production by Klebsiella pneumoniae isolated from human tongue surface.
Sensors (Basel, Switzerland) (2012-06-28)
Wai-Fong Yin, Kathiravan Purmal, Shenyang Chin, Xin-Yue Chan, Chong-Lek Koh, Choon-Kook Sam, Kok-Gan Chan
RESUMEN
Bacteria communicate by producing quorum sensing molecules called autoinducers, which include autoinducer-1, an N-hexanoyl homoserine lactone (AHL), and autoinducer-2. Bacteria present in the human oral cavity have been shown to produce autoinducer-2, but not AHL. Here, we report the isolation of two AHL-producing Klebsiella pneumoniae strains from the posterior dorsal surface of the tongue of a healthy individual. Spent culture supernatant extracts from K. pneumoniae activated the biosensors Agrobacterium tumefaciens NTL4(pZLR4) and Escherichia coli [pSB401], suggesting the presence of both long and short chain AHLs. High resolution mass spectrometry analyses of these extracts confirmed that both K. pneumoniae isolates produced N-octanoylhomoserine lactone and N-3-dodecanoyl-L-homoserine lactone. To the best of our knowledge, this is the first report of the isolation of K. pneumoniae from the posterior dorsal surface of the human tongue and the production of these AHLs by this bacterium.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
N-Heptanoyl-L-homoserine lactone, ≥96% (HPLC)