Merck
  • Candida albicans Czf1 and Efg1 coordinate the response to farnesol during quorum sensing, white-opaque thermal dimorphism, and cell death.
Candida albicans Czf1 and Efg1 coordinate the response to farnesol during quorum sensing, white-opaque thermal dimorphism, and cell death.
Eukaryotic cell (2013-07-23)
Melanie L Langford, Jessica C Hargarten, Krista D Patefield, Elizabeth Marta, Jill R Blankenship, Saranna Fanning, Kenneth W Nickerson, Audrey L Atkin
RESUMEN
Quorum sensing by farnesol in Candida albicans inhibits filamentation and may be directly related to its ability to cause both mucosal and systemic diseases. The Ras1-cyclic AMP signaling pathway is a target for farnesol inhibition. However, a clear understanding of the downstream effectors of the morphological farnesol response has yet to be unraveled. To address this issue, we screened a library for mutants that fail to respond to farnesol. Six mutants were identified, and the czf1Δ/czf1Δ mutant was selected for further characterization. Czf1 is a transcription factor that regulates filamentation in embedded agar and also white-to-opaque switching. We found that Czf1 is required for filament inhibition by farnesol under at least three distinct environmental conditions: on agar surfaces, in liquid medium, and when embedded in a semisolid agar matrix. Since Efg1 is a transcription factor of the Ras1-cyclic AMP signaling pathway that interacts with and regulates Czf1, an efg1Δ/efg1Δ czf1Δ/czf1Δ mutant was tested for filament inhibition by farnesol. It exhibited an opaque-cell-like temperature-dependent morphology, and it was killed by low farnesol levels that are sublethal to wild-type cells and both efg1Δ/efg1Δ and czf1Δ/czf1Δ single mutants. These results highlight a new role for Czf1 as a downstream effector of the morphological response to farnesol, and along with Efg1, Czf1 is involved in the control of farnesol-mediated cell death in C. albicans.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Farnesol, 95%
Sigma-Aldrich
Farnesol, mixture of isomers, ≥95%, stabilized, FG