Merck
Cerebrospinal fluid Aβ42, phosphorylated Tau181, and resting-state functional connectivity.
JAMA neurology (2013-08-21)
Liang Wang, Matthew R Brier, Abraham Z Snyder, Jewell B Thomas, Anne M Fagan, Chengjie Xiong, Tammie L Benzinger, David M Holtzman, John C Morris, Beau M Ances
RESUMEN
Resting-state functional connectivity magnetic resonance imaging has great potential for characterizing pathophysiological changes during the preclinical phase of Alzheimer disease. To assess the relationship between default mode network integrity and cerebrospinal fluid biomarkers of Alzheimer disease pathology in cognitively normal older individuals. Cross-sectional cohort study at The Charles F. and Joanne Knight Alzheimer's Disease Research Center at Washington University in St Louis, St Louis, Missouri, among 207 older adults with normal cognition (Clinical Dementia Rating, 0). Resting-state functional connectivity magnetic resonance imaging measures of default mode network integrity. Decreased cerebrospinal fluid Aβ42 and increased cerebrospinal fluid phosphorylated tau181 were independently associated with reduced default mode network integrity, with the most prominent decreases in functional connectivity observed between the posterior cingulate and medial temporal regions. Observed reductions in functional connectivity were unattributable to age or structural atrophy in the posterior cingulate and medial temporal areas. Similar resting-state functional connectivity magnetic resonance imaging findings in relation to cerebrospinal fluid biomarkers were obtained using region-of-interest analyses and voxelwise correlation mapping. Both Aβ and tau pathology affect default mode network integrity before clinical onset of Alzheimer disease.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
L-Threonine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
L-Threonine, reagent grade, ≥98% (HPLC)
SAFC
L-Threonine
Sigma-Aldrich
L-Threonine, BioXtra, ≥99.5% (NT)
Supelco
L-Threonine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Threonine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland