Merck
Traditional and novel bone remodeling markers in premenopausal and postmenopausal women.
The Journal of clinical endocrinology and metabolism (2013-09-05)
Sonsoles Botella, Patricia Restituto, Ignacio Monreal, Inmaculada Colina, Amparo Calleja, Nerea Varo
RESUMEN
Bone turnover markers (BTMs) may identify changes in bone remodeling within a relatively short time interval before changes in bone mineral density can be detected. New markers such as osteoprotegerin, receptor activator of nuclear factor-κB ligand, and sclerostin have emerged, but there is little information about their potential use in clinical practice. The aim of this study was to analyze the ability of several BTMs to predict bone loss in pre- and postmenopausal women and to monitor the efficacy of treatment in osteoporotic women. We performed an observational prospective study in pre- and postmenopausal ambulatory women (n = 72 and n = 152, respectively). Postmenopausal women with osteoporosis (n = 18) were treated with risedronate and calcium. Women filled out a questionnaire and underwent bone mineral density measurement using dual-energy x-ray absorptiometry at the time of enrollment and after 1 year of follow-up. BTMs were measured at baseline, at 6 months, and after 1 year. Increased levels of N-terminal propeptide of type 1 procollagen (P1NP) and β-type I collagen telopeptides (CTXs) were associated with low bone mineral density in the premenopausal (P = .02 and P = .04, respectively) and postmenopausal (P = .03 and P = .02) groups. The best analytical performance to diagnose osteoporosis was for β-CTX, osteocalcin, and P1NP, with areas under the curve of 0.70 (P = .005), 0.64 (P = .048), and 0.71 (P = .003). A significant decrease was found in P1NP, osteocalcin, tartrate-resistant acid phosphatase-5b, β-CTX, and bone alkaline phosphatase after 1 year of treatment (all P < .05). Our data suggest that measurement of β-CTX and P1NP shows adequate analytical performance and could potentially be included in algorithms for the screening of osteoporosis. Furthermore, these two markers, along with osteocalcin and tartrate-resistant acid phosphatase-5b, are useful to monitor the response to risedronate.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥0.5 unit/mg solid
Sigma-Aldrich
Phosphatase, Acid from wheat germ, ≥0.4 unit/mg solid
Sigma-Aldrich
Phosphatase, Acid from potato, lyophilized powder, ≥3.0 units/mg solid
Sigma-Aldrich
Phosphatase, Acid from sweet potato, ammonium sulfate suspension, ≥10.0 units/mg protein (modified Warburg-Christian)