Merck
Identification of differentially expressed proteins in fresh and frozen-thawed boar spermatozoa by iTRAQ-coupled 2D LC-MS/MS.
Reproduction (Cambridge, England) (2013-12-21)
Xiaoli Chen, Huabin Zhu, Chuanhuo Hu, Haisheng Hao, Junfang Zhang, Kunpeng Li, Xueming Zhao, Tong Qin, Kan Zhao, Huishan Zhu, Dong Wang
RESUMEN
Cryodamage is a major problem in semen cryopreservation, causing changes in the levels of proteins that influence the function and motility of spermatozoa. In this study, protein samples prepared from fresh and frozen-thawed boar spermatozoa were compared using the isobaric tags for relative and absolute quantification (iTRAQ) labeling technique coupled to 2D LC-MS/MS analysis. A total of 41 differentially expressed proteins were identified and quantified, including 35 proteins that were present at higher levels and six proteins that were present at lower levels in frozen-thawed spermatozoa by at least a mean of 1.79-fold (P<0.05). On classifying into ten distinct categories using bioinformatic analysis, most of the 41 differentially expressed proteins were found to be closely relevant to sperm premature capacitation, adhesions, energy supply, and sperm-oocyte binding and fusion. The expression of four of these proteins, SOD1, TPI1, ODF2, and AKAP3, was verified by western blot analysis. We propose that alterations in these identified proteins affect the quality of cryopreserved semen and ultimately lower its fertilizing capacity. This is the first study to compare protein levels in fresh and frozen-thawed spermatozoa using the iTRAQ technology. Our preliminary results provide an overview of the molecular mechanisms of cryodamage in frozen-thawed spermatozoa and theoretical guidance to improve the cryopreservation of boar semen.