Merck
[Analysis of penbutolol and its metabolites in human body fluid by gas chromatography--mass spectrometry].
Yao xue xue bao = Acta pharmaceutica Sinica (1995-01-01)
L Shen, Y X Xu, C J Zhang, Y Wu, S Wang, K R Cui
RESUMEN
A method for the detection and identification of penbutolol and its metabolites in human body fluid by using gas chromatography-mass spectrometry was established. The urine was extracted with diethyl ether-isopropanol and the plasma was extracted with Sep-Pak column. After TMS derivatization the sample was analyzed with GC-MSD. Six metabolites of penbutolol were found in urine sample. The recovery was 90.83% for urine and 85.88% for plasma. The detection limit of penbutolol was 5 pg.
MATERIALES
Número de producto
Marca
Descripción del producto
Penbutolol sulfate, European Pharmacopoeia (EP) Reference Standard