Merck
Oxaprozin: a new NSAID.
Orthopaedic review (1992-05-01)
M A al-Faks, M C Pugh
RESUMEN
Oxaprozin, a propionic acid derivative, is a non-steroidal anti-inflammatory drug (NSAID) with analgesic and antipyretic properties. Its spectrum of anti-inflammatory activity has been demonstrated in both in vitro and in vivo models.1 The majority of published clinical trials have focused on the use of oxaprozin (from Searle) in the treatment of osteoarthritis and rheumatoid arthritis.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Oxaprozin, solid
En este momento no podemos mostrarle ni los precios ni la disponibilidad
USP
Oxaprozin, United States Pharmacopeia (USP) Reference Standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad