Merck
Caffeic Acid, a versatile pharmacophore: an overview.
Mini reviews in medicinal chemistry (2011-06-18)
M Touaibia, J Jean-François, J Doiron
RESUMEN
The caffeic acid scaffold, which is abundant in nature, is extremely versatile and is found in a number of biologically active molecules. The purpose of this review is to provide an overview of the pharmacological activity of synthetic caffeic acid analogs including recent reports of anti-inflammatory, anti-cancer, and antiviral activities of these compounds.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Caffeic acid, ≥98.0% (HPLC)
Supelco
Caffeic acid, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)