Merck
Swainsonine: a new antineoplastic immunomodulator.
Journal of the National Medical Association (1989-10-01)
S Mohla, M J Humphries, S L White, K Matsumoto, S A Newton, C C Sampson, D Bowen, K Olden
RESUMEN
Swainsonine, an indolizidine alkaloid with immunomodulatory activity, has been found to be effective in inhibiting metastatic dissemination and growth of primary tumors of both murine and human origins. The unique ability of swainsonine to exhibit antimetastatic, anti-proliferative, and immunomodulatory activity imparts this drug a promising future in cancer therapy.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Swainsonine, synthetic
Sigma-Aldrich
Swainsonine, from Metarrhizium anisopliae, ≥98% (TLC)