Merck
Mutations of the RDX gene cause nonsyndromic hearing loss at the DFNB24 locus.
Human mutation (2007-01-18)
Shahid Y Khan, Zubair M Ahmed, Muhammad I Shabbir, Shin-ichiro Kitajiri, Saeeda Kalsoom, Saba Tasneem, Sara Shayiq, Arabandi Ramesh, Srikumari Srisailpathy, Shaheen N Khan, Richard J H Smith, Saima Riazuddin, Thomas B Friedman, Sheikh Riazuddin
RESUMEN
Ezrin, radixin, and moesin are paralogous proteins that make up the ERM family and function as cross-linkers between integral membrane proteins and actin filaments of the cytoskeleton. In the mouse, a null allele of Rdx encoding radixin is associated with hearing loss as a result of the degeneration of inner ear hair cells as well as with hyperbilirubinemia due to hepatocyte dysfunction. Two mutant alleles of RDX [c.1732G>A (p.D578N) and c.1404_1405insG (p.A469fsX487)] segregating in two consanguineous Pakistani families are associated with neurosensory hearing loss. Both of these mutant alleles are predicted to affect the actin-binding motif of radixin. Sequence analysis of RDX in the DNA samples from the original DFNB24 family revealed a c.463C>T transition substitution that is predicted to truncate the protein in the FERM domain (F for 4.1, E for ezrin, R for radixin, and M for moesin) (p.Q155X). We also report a more complete gene and protein structure of RDX, including four additional exons and five new isoforms of RDX that are expressed in human retina and inner ear. Further, high-resolution confocal microscopy in mouse inner ear demonstrates that radixin is expressed along the length of stereocilia of hair cells from both the organ of Corti and the vestibular system.