Merck
Pnpla3I148M knockin mice accumulate PNPLA3 on lipid droplets and develop hepatic steatosis.
Hepatology (Baltimore, Md.) (2014-06-12)
Eriks Smagris, Soumik BasuRay, John Li, Yongcheng Huang, Ka-man V Lai, Jesper Gromada, Jonathan C Cohen, Helen H Hobbs
RESUMEN
A sequence polymorphism (rs738409, I148M) in patatin-like phospholipid domain containing protein 3 (PNPLA3) is strongly associated with nonalcoholic fatty liver disease (NAFLD), but the mechanistic basis for this association remains enigmatic. Neither ablation nor overexpression of wild-type PNPLA3 affects liver fat content in mice, whereas hepatic overexpression of the human 148M transgene causes steatosis. To determine whether the 148M allele causes fat accumulation in the liver when expressed at physiological levels, we introduced a methionine codon at position 148 of the mouse Pnpla3 gene. Knockin mice had normal levels of hepatic fat on a chow diet, but when challenged with a high-sucrose diet their liver fat levels increased 2 to 3-fold compared to wild-type littermates without any associated changes in glucose homeostasis. The increased liver fat in the knockin mice was accompanied by a 40-fold increase in PNPLA3 on hepatic lipid droplets, with no increase in hepatic PNPLA3 messenger RNA (mRNA). Similar results were obtained when the catalytic dyad of PNPLA3 was inactivated by substituting the catalytic serine with alanine (S47A). These data provide the first direct evidence that physiological expression of PNPLA3 148M variant causes NAFLD, and that the accumulation of catalytically inactive PNPLA3 on the surfaces of lipid droplets is associated with the accumulation of TG in the liver.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Sacarosa, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
Sigma-Aldrich
Colesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sacarosa, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Supelco
Sacarosa, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Sacarosa, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Sacarosa, ≥99.5% (GC)
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Colesterol, powder, BioReagent, suitable for cell culture, ≥99%
Millipore
Sacarosa, suitable for microbiology, ACS reagent, ≥99.0%
Sigma-Aldrich
Sacarosa, ACS reagent
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
Sacarosa, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Colesterol, from sheep wool, ≥92.5% (GC), powder
Supelco
Colesterol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sacarosa, puriss., meets analytical specification of Ph. Eur., BP, NF
Supelco
Cholesterol solution, certified reference material, 10 mg/mL in chloroform
Sigma-Aldrich
Sacarosa, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sacarosa, meets USP testing specifications
Supelco
Sacarosa, analytical standard, for enzymatic assay kit SCA20
Sigma-Aldrich
Colesterol, from lanolin, ≥99.0% (GC)
SAFC
Colesterol, from sheep wool, Controlled origin, meets USP/NF testing specifications
Sigma-Aldrich
Colesterol, tested according to Ph. Eur.