Merck
  • Duodenum-triggered delivery of pravastatin sodium via enteric surface-coated nanovesicular spanlastic dispersions: development, characterization and pharmacokinetic assessments.
Duodenum-triggered delivery of pravastatin sodium via enteric surface-coated nanovesicular spanlastic dispersions: development, characterization and pharmacokinetic assessments.
International journal of pharmaceutics (2015-02-11)
Saadia Ahmed Tayel, Mohamed Ahmed El-Nabarawi, Mina Ibrahim Tadros, Wessam Hamdy Abd-Elsalam
RESUMEN
Pravastatin sodium (PVS) is a hydrophilic HMG-CoA reductase inhibitor that is mainly absorbed from duodenum. PVS has a short elimination half-life (1-3 h), suffers from instability at gastric pH, extensive hepatic first-pass metabolism and low absolute bioavailability (18%). The current work aimed to develop enteric surface-coated spanlastic dispersions as controlled-release duodenum-triggered systems able to surmount PVS drawbacks. PVS-loaded spanlastic dispersions were prepared by ethanol-injection method using span(®) 60. Tween(®) 60 and Tween(®) 80 were explored as edge activators. As a novel approach, the fine spanlastic dispersions were surface-coated with an enteric-polymer (Eudragit(®) L100-55) via freeze-drying. The systems were evaluated, before and after enteric-coating, for particle size, zeta potential, PVS entrapment efficiency (EE%), morphology and PVS release studies. PVS pharmacokinetics from the best achieved system and an aqueous solution were estimated in rats by UPLC-MS/MS. The best achieved enteric surface-coated spanlastic dispersion (E-S6) displayed spherical nanosized vesicles (647.60 nm) possessing negative zeta potential (-6.93 mV), promising EE% (63.22%) and a biphasic drug-release pattern characterized by a retarded-release phase (0.1 N HCl, 2 h) and a controlled-release phase (pH 6.8, 10 h). The higher Cmax, delayed Tmax, prolonged MRT(0-∞), longer elimination t50% and enhanced oral bioavailability unravel E-S6 potential for oral PVS delivery.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Metanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Fosfato de potasio monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Fosfato de sodio dibasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Ácido fórmico, reagent grade, ≥95%
Sigma-Aldrich
Ácido fosfórico, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Metanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥96%
Sigma-Aldrich
Ácido fosfórico, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Metanol, anhydrous, 99.8%
Sigma-Aldrich
Ácido fosfórico, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Fosfato de sodio dibasic, puriss. p.a., ACS reagent, anhydrous, ≥99.0% (T)
Sigma-Aldrich
Fosfato de potasio monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Metanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Fosfato de sodio dibasic, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E 339, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Metanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Fosfato de potasio monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Ácido fórmico, ACS reagent, ≥88%
Sigma-Aldrich
Metanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ácido fosfórico, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Ácido fosfórico, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Fosfato de potasio monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Ácido fosfórico, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Fosfato de sodio dibasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ácido fosfórico, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Fosfato de potasio monobasic, ReagentPlus®
Sigma-Aldrich
Metanol, Absolute - Acetone free