Merck
The iron content and ferritin contribution in fresh, dried, and toasted nori, Pyropia yezoensis.
Bioscience, biotechnology, and biochemistry (2014-10-16)
Taro Masuda, Ami Yamamoto, Haruhiko Toyohara
RESUMEN
Iron is one of the essential trace elements for humans. In this study, the iron contents in fresh, dried, and toasted nori (Pyropia yezoensis) were analyzed. The mean iron content of fresh, dried, and toasted nori were 19.0, 22.6, and 26.2 mg/100 g (dry weight), respectively. These values were superior to other food of plant origin. Furthermore, most of the iron in nori was maintained during processing, such as washing, drying, and toasting. Then, the form of iron in fresh, dried, and toasted nori was analyzed. As a result, an iron storage protein ferritin contributed to iron storage in raw and dried nori, although the precise rate of its contribution is yet to be determined, while ferritin protein cage was degraded in the toasted nori. It is the first report that verified the ferritin contribution to iron storage in such edible macroalgae with commercial importance.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido nítrico, ACS reagent, 70%
Sigma-Aldrich
Sulfato de amonio, ACS reagent, ≥99.0%
Sigma-Aldrich
Ácido nítrico, 70%, purified by redistillation, ≥99.999% trace metals basis
Sigma-Aldrich
Sulfato de amonio, Molecular Biology, ≥99.0%
Sigma-Aldrich
Sulfato de amonio, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ácido nítrico, puriss. p.a., 65.0-67.0%
Millipore
Peróxido de hidrógeno solution, 3%, suitable for microbiology
Sigma-Aldrich
Peróxido de hidrógeno solution, contains ~200 ppm acetanilide as stabilizer, 3 wt. % in H2O
Sigma-Aldrich
Ácido nítrico, puriss. p.a., reag. ISO, reag. Ph. Eur., ≥65%
Sigma-Aldrich
Ácido nítrico, puriss. p.a., ≥65% (T)
Supelco
Ácido nítrico solution, 0.1 M HNO3 in water (0.1N), eluent concentrate for IC
Sigma-Aldrich
Ácido nítrico, ACS reagent, ≥90.0%
Sigma-Aldrich
Sulfato de amonio, BioXtra, ≥99.0%
Sigma-Aldrich
Sulfato de amonio, BioUltra, ≥99.0% (T)
Supelco
Sulfato de amonio, analytical standard, for Nitrogen Determination According to Kjeldahl Method, traceable to NIST SRM 194
Sigma-Aldrich
Sulfato de amonio, 99.999% trace metals basis
Sigma-Aldrich
Sulfato de amonio, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sulfato de amonio, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sulfato de amonio, anhydrous, Redi-Dri, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium-14N2 sulfate, 99.99 atom % 14N
Sigma-Aldrich
Nitric-14N acid solution, ~10 N in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2 sulfate solution, 40 wt. % in H2O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium-14N2,sulfate-16O4, 99.99 atom % 16O, 99.99 atom % 14N
Sigma-Aldrich
Ammonium sulfate-14N2 solution, 40 wt. % in H2O, 99.99 atom % 14N