Merck
Rapid determination of lamivudine in human plasma by high-performance liquid chromatography.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-02)
Mahmoud Alebouyeh, Hossein Amini
RESUMEN
A simple and rapid high-performance liquid chromatographic method with spectrophotometric detection was developed for the determination of lamivudine in human plasma. Sample preparation was accomplished through protein precipitation with acetonitrile followed by aqueous phase separation using dichloromethane. Lamivudine and the internal standard acyclovir were well separated from endogenous plasma peaks on a Chromolith RP-18e column under isocratic elution with 50 mM sodium dihydrogen phosphate-triethylamine (996:4, v/v), pH 3.2 at 20 °C. Total run time at a flow-rate of 1.5 ml/min was less than 5 min. Detection was made at 278 nm. The method was specific and sensitive, with a lower quantification limit of 40 ng/ml and a detection limit of 10 ng/ml. The absolute recovery was 97.7%, while the within- and between-day coefficient of variation and percent error values of the assay method were all less than 7%. The linearity was assessed in the range of 40-2560 in plasma, with a correlation coefficient of greater than 0.999. The method was successfully applied to a bioequivalence study in healthy volunteers.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Diclorometano, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Diclorometano, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Ácido fosfórico, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Diclorometano, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Diclorometano, anhydrous, ≥99.8%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ácido fosfórico, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Ácido fosfórico, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Sodium phosphate monobasic, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Diclorometano, puriss. p.a., ACS reagent, reag. ISO, ≥99.9% (GC)
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ácido fosfórico, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥85%
Sigma-Aldrich
Sodium phosphate monobasic, BioReagent, Molecular Biology, anhydrous, ≥98%
Sigma-Aldrich
Ácido fosfórico, puriss. p.a., crystallized, ≥99.0% (T)
Sigma-Aldrich
Ácido fosfórico, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Ácido fosfórico, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, 85.0-88.0%
Sigma-Aldrich
Sodium phosphate monobasic, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0% (titration)
Sigma-Aldrich
Diclorometano, puriss., meets analytical specification of Ph. Eur., NF, ≥99% (GC)
Sigma-Aldrich
Ácido fosfórico, 85 wt. % in H2O, FCC, FG
Supelco
Diclorometano, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ácido fosfórico, BioUltra, ≥85% (T)
Sigma-Aldrich
Sodium phosphate monobasic, meets USP testing specifications, anhydrous
Sigma-Aldrich
Sodium phosphate monobasic, purum p.a., anhydrous, ≥99.0% (T)
Sigma-Aldrich
Sodium phosphate monobasic solution, BioUltra, 5 M in H2O
Sigma-Aldrich
Sodium phosphate monobasic, BioXtra, ≥99.0%
Supelco
Diclorometano, analytical standard
Sigma-Aldrich
Sodium phosphate monobasic, anhydrous, free-flowing, Redi-Dri, ≥99.0%
Sigma-Aldrich
Diclorometano, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Ácido fosfórico, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Supelco
Acyclovir, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ácido fosfórico, BioReagent, suitable for insect cell culture, 85%