Merck
Plasma-spray ionization (PLASI): a multimodal atmospheric pressure ion source for liquid stream analysis.
Journal of the American Society for Mass Spectrometry (2014-07-09)
Adam Kaylor, Prabha Dwivedi, Jennifer J Pittman, María Eugenia Monge, Guilong Cheng, Shelly Li, Facundo M Fernández
RESUMEN
A new ion generation method, named plasma-spray ionization (PLASI) for direct analysis of liquid streams, such as in continuous infusion experiments or liquid chromatography (LC), is reported. PLASI addresses many of the analytical limitations of electrospray ionization (ESI) and has potential for real time process stream analysis and reaction monitoring under atmospheric conditions in non-ESI friendly scenarios. In PLASI-mass spectrometry (MS), the liquid stream is pneumatically nebulized and partially charged at low voltages; the resultant aerosol is thus entrained with a gaseous plasma plume from a distal glow discharge prior to MS detection. PLASI-MS not only overcomes ESI-MS limitations but also generates simpler mass spectra with minimal adduct and cluster formation. PLASI utilizes the atomization capabilities of an ESI sprayer operated below the ESI threshold to generate gas-phase aerosols that are then ionized by the plasma stream. When operated at or above the ESI threshold, ionization by traditional ESI mechanisms is achieved. The multimodal nature of the technique enables readily switching between plasma and ESI operation. It is expected that PLASI will enable analyzing a wide range of analytes in complex matrices and less-restricted solvent systems, providing more flexibility than that achievable by ESI alone.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Metanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Fosfato de potasio monobasic, ACS reagent, ≥99.0%
Sigma-Aldrich
Metanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Metanol, anhydrous, 99.8%
Sigma-Aldrich
Fosfato de potasio monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Supelco
Caffeine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Metil 4-hidroxibenzoato, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Metanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Metanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Caffeine, powder, ReagentPlus®
Sigma-Aldrich
Fosfato de potasio monobasic, meets analytical specification of Ph. Eur., NF, E340, anhydrous, 98-100.5% (calc. to the dried substance)
Sigma-Aldrich
Metanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Fosfato de potasio monobasic, buffer substance, anhydrous, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 99.5-100.5%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Fosfato de potasio monobasic, ReagentPlus®
Sigma-Aldrich
Metanol, Absolute - Acetone free
Sigma-Aldrich
Metil 4-hidroxibenzoato, ReagentPlus®, ≥99.0%, crystalline
Sigma-Aldrich
Fosfato de potasio monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Caffeine, anhydrous, 99%, FCC, FG
USP
Metanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Metanol, BioReagent, ≥99.93%
Supelco
Metanol, analytical standard
USP
Metilparabeno, United States Pharmacopeia (USP) Reference Standard
Supelco
Metanol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Caffeine, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Supelco
Schmelzpunktstandard 235-237°C, analytical standard