Merck
PDE5 inhibitors enhance the lethality of standard of care chemotherapy in pediatric CNS tumor cells.
Cancer biology & therapy (2014-03-22)
Jane L Roberts, Laurence Booth, Adam Conley, Nichola Cruickshanks, Mark Malkin, Rakesh C Kukreja, Steven Grant, Andrew Poklepovic, Paul Dent
RESUMEN
We determined whether clinically relevant phosphodiesterase 5 (PDE5) inhibitors interacted with clinically relevant chemotherapies to kill medulloblastoma cells. In medulloblastoma cells PDE5 inhibitors interacted in a greater than additive fashion with vincristine/etoposide/cisplatin to cause cell death. Knockdown of PDE5 expression recapitulated the combination effects of PDE5 inhibitor drugs with chemotherapy drugs. Expression of dominant negative caspase 9 did not significantly inhibit chemotherapy lethality but did significantly reduce enhanced killing in combination with the PDE5 inhibitor sildenafil. Overexpression of BCL-XL and c-FLIP-s suppressed individual and combination drug toxicities. Knockdown of CD95 or FADD suppressed drug combination toxicity. Treatment with PDE5 inhibitors and chemotherapy drugs promoted autophagy which was maximal at ~12 h post-treatment, and in a cell type-dependent manner knockdown of Beclin1 or ATG5 either suppressed or enhanced drug combination lethality. PDE5 inhibitors enhanced the induction of chemotherapy-induced DNA damage in a nitric oxide synthase-dependent fashion. In conclusion, our data demonstrate that the combination of PDE5 inhibitors with standard of care chemotherapy agents for medulloblastoma represents a possible novel modality for future treatment of this disease.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
MISSION® esiRNA, targeting human GAPDH
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Gapdh
En este momento no podemos mostrarle ni los precios ni la disponibilidad