Merck
  • Reconstituted mother tinctures of Gelsemium sempervirens L. improve memory and cognitive impairment in mice scopolamine-induced dementia model.
Reconstituted mother tinctures of Gelsemium sempervirens L. improve memory and cognitive impairment in mice scopolamine-induced dementia model.
Journal of ethnopharmacology (2014-12-03)
Partha Palit, Dhrubojyoti Mukherjee, Subhash C Mandal
RESUMEN
Gelsemium sempervirens (L.) J.St.-Hil is a herb used for the treatment of various neuroses in both homeopathic and Ayurvedic systems. The present study examines whether Gelsemium reconstituted tincture can protect against scopolamine induced cognitive discrepancies in amnesic mouse model. In order to investigate the protective mechanism of Gelsemium against dementia, in vitro acetyl cholinesterase and β-secretase enzyme inhibition and estimation of glutathione level in mouse brain were carried out. The inhibition study on acetyl cholinesterase and β-secretase enzyme was conducted on brain homogenate supernatant spectrophotometrically using specific substrate. Cognitive enhancement activity was assessed by elevated plus maze and passive avoidance study in scopolamine induced dementia mouse model. Glutathione, an anti-oxidant, was measured spectrophotometrically from scopolamine induced amnesic mice brain supernatant using 5,5'-dithiobis 2-nitrobenzoic acid in the presence and absence of Gelsemium tincture. Significant inhibition was found with Gelsemium on AChE and β-secretase enzyme with an IC50 of 9.25 and 16.25 µg/ml, respectively, followed by increasing glutathione levels in comparison to the untreated dementia group. The effect of Gelsemium of scopolamine-induced cognitive deficits was determined by measuring the behavioral parameters and the antioxidant status of the brain after scopolamine (1mg/kg i.p.) injected amnesic mice. Gelsemium significantly demonstrated in vivo anti-dementia activity (60% protection) and increased exploratory behavior. Our investigations indicated that alkaloid, iridoids and coumarin enriched reconstituted Gelsemium tincture extract displays promising cognitive enhancement in adult mice after short-term oral treatment. Hence, Gelsemium can be a promising anti-dementia agent, mediating the protection against amnesia, attention disorders and learning dysfunctions through dual inhibition of both acetyl cholinesterases (no false positive effect was shown), β-secretase and antioxidant activity.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido acético, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Ácido acético, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Ammonium bicarbonate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Ácido acético, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Ácido acético solution, suitable for HPLC
Sigma-Aldrich
Ácido acético, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Ácido acético, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, anhydrous, Molecular Biology, ≥99%
Sigma-Aldrich
Ammonium bicarbonate, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Acetylthiocholine iodide, ≥98% (TLC), powder or crystals
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Ácido acético, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate, >99%, FG
USP
Ácido acético, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetylthiocholine iodide, ≥99.0% (AT)
Sigma-Aldrich
Sodium acetate, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Supelco
Ácido acético, analytical standard
Sigma-Aldrich
Sodium acetate solution, BioUltra, Molecular Biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, suitable for luminescence, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Ácido acético, ≥99.5%, FCC, FG
Sigma-Aldrich
Piracetam
Sigma-Aldrich
Ácido acético, natural, ≥99.5%, FG
Sigma-Aldrich
Ácido acético, glacial, puriss., 99-100%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
2-Nitrobenzoic acid, 95%, Contains 3- and 4-isomers
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%