Merck
Histamine monolith versatility to purify supercoiled plasmid deoxyribonucleic acid from Escherichia coli lysate.
Journal of chromatography. A (2014-06-21)
A Sousa, A M Almeida, U Černigoj, F Sousa, J A Queiroz
RESUMEN
Preparation of high quantities of supercoiled plasmid DNA of pharmaceutical grade purity is a research area where intensive investigation is being performed. From this standpoint, several downstream methods have been proposed, among them the monolithic chromatographic strategies owing to excellent mass transfer properties of monolithic supports and their high binding capacity for large biomolecules. The present study explores the physicochemical properties of histamine ligand in a supercoiled plasmid DNA purification process from an Escherichia coli clarified lysate, where the emphasis is given to the elution strategy that allows higher selectivity and efficient removal of other impurities besides the open circular isoform. The combination of high NaCl concentration and acidic pH allowed the elimination of 89% of RNA during the preparative loading of the lysate sample. The results of the purification strategy with ascending sodium chloride gradient revealed that 97% of supercoiled plasmid DNA was recovered with a purity degree of 99%. In addition, using a combined purification strategy with ascending sodium chloride (capture step) and then descending ammonium sulfate (polishing step) gradient, it was achieved a lower supercoiled plasmid DNA recovery yield of 79% with a purity degree of 92%, although the dynamic binding capacity under these conditions was higher than in the previous strategy. A significant reduction of host contents, such as proteins, RNA and genomic DNA, was obtained in both purification strategies. Accordingly, histamine is a useful and versatile ligand that allows the desirable supercoiled plasmid purification with high yield and purity level.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Base Trizma®, Primary Standard and Buffer, ≥99.9% (titration), crystalline
Sigma-Aldrich
Sulfato de amonio, ACS reagent, ≥99.0%
Sigma-Aldrich
Base Trizma®, BioPerformance Certified, meets EP, USP testing specifications, suitable for cell culture, ≥99.9% (titration)
Sigma-Aldrich
Ácido cítrico, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Cloruro de sodio, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sulfato de amonio, Molecular Biology, ≥99.0%
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ácido cítrico, ACS reagent, ≥99.5%
Sigma-Aldrich
Tris(hidroximetil)-aminometano, ACS reagent, ≥99.8%
Sigma-Aldrich
Tromethamine, meets USP testing specifications
Sigma-Aldrich
Sulfato de amonio, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sigma 7-9®, ≥99% (titration), powder or crystals
Sigma-Aldrich
Base Trizma®, BioUltra, Molecular Biology, ≥99.8% (T)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Ácido cítrico, ≥99.5%, FCC, FG
Sigma-Aldrich
Ácido cítrico, 99%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Ácido cítrico, anhydrous, suitable for cell culture, suitable for plant cell culture
Sigma-Aldrich
Cloruro de sodio, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Histamine, ≥97.0%
Sigma-Aldrich
Base Trizma®, ≥99.0% (T)
Sigma-Aldrich
Cloruro de sodio, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Cloruro de sodio, 99.999% trace metals basis
Supelco
Cloruro de sodio, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Ácido cítrico, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Base Trizma®, BioXtra, pH 10.5-12.0 (1 M in H2O), ≥99.9% (titration)