Merck
Towards Elucidating the Effects of Purified MWCNTs on Human Lung Epithelial cells.
Environmental science. Nano (2014-12-09)
Chenbo Dong, Reem EIdawud, Linda M Sargent, Michael L Kashon, David Lowry, Yon Rojanasakul, Cerasela Zoica Dinu
RESUMEN
Toxicity of engineered nanomaterials is associated with their inherent properties, both physical and chemical. Recent studies have shown that exposure to multi-walled carbon nanotubes (MWCNTs) promotes tumors and tumor-associated pathologies and lead to carcinogenesis in model
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Alcohol etílico puro 200, Molecular Biology
Sigma-Aldrich
Alcohol etílico puro, 200 proof, ACS reagent, ≥99.5%
Sigma-Aldrich
Alcohol etílico puro, 200 proof, meets USP testing specifications
Sigma-Aldrich
1,2-Dicloroetano, ACS reagent, ≥99.0%
Sigma-Aldrich
Alcohol etílico puro, 200 proof, anhydrous, ≥99.5%
Sigma-Aldrich
Alcohol etílico puro 190, for molecular biology
Sigma-Aldrich
N-Hydroxysuccinimide, 98%
Sigma-Aldrich
L-Glutamina, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Yoduro de propidio, ≥94.0% (HPLC)
Sigma-Aldrich
Alcohol etílico puro, 190 proof, ACS spectrophotometric grade, 95.0%
Sigma-Aldrich
1,2-Dicloroetano, anhydrous, 99.8%
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, commercial grade, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
Sigma-Aldrich
L-Glutamina, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, ≥99.0% (AT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, crystalline
SAFC
L-Glutamina
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, purum, ≥98.0% (AT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Supelco
Etanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
N-(3-dimetilaminopropilo)-N′-etilcarbodiimida hydrochloride, BioXtra
Supelco
Etanol, standard for GC
Sigma-Aldrich
Propidium iodide solution