Merck
Effects of glutamine and asparagine on recombinant antibody production using CHO-GS cell lines.
Biotechnology progress (2014-08-01)
Ping Xu, Xiao-Ping Dai, Erica Graf, Richard Martel, Reb Russell
RESUMEN
A unique and nontraditional approach using glutamine and asparagine supplements for CHO-glutamine synthetase (GS) cell lines was studied. In our experiments, we found that a decrease in pH and an increase in cell death occurred in production phase of a GS cell line, leading to reduced antibody expression and lower antibody yields. The experimental results and the statistical analysis (ANOVA) indicated that additions of glutamine and asparagine in the basal and feed media were effective to buffer the cell culture pH, reduce lactate generation, maintain a higher cell viability profile, and improve antibody productivity. In bench-top bioreactors, glutamine and asparagine supplementation helped to prevent cell death, improve antibody yield, and reduce base usage. Glutamine is normally excluded from culture media for GS cell lines to prevent the bypass of selection pressure. In this study, however, the addition of glutamine did not affect cell population homogeneity, protein quality, or decrease antibody yield of two GS cell lines.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Metanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Metanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Formaldehído solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Metanol, anhydrous, 99.8%
Sigma-Aldrich
Ammonia solution, 7 N in methanol
Sigma-Aldrich
Formaldehído solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehído solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Metanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Metanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Metanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanol, Absolute - Acetone free
Sigma-Aldrich
Formaldehído solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Ammonia solution, 2.0 M in ethanol
USP
Metanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Metanol, BioReagent, ≥99.93%
Supelco
Metanol, analytical standard
Sigma-Aldrich
Ammonia solution, 0.4 M in dioxane
Supelco
Metanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonia solution, 4 M in methanol
Sigma-Aldrich
Ammonia solution, 0.4 M in THF
Sigma-Aldrich
Formaldehído solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Supelco
Formaldehído solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Ammonia solution, 2.0 M in methanol
Sigma-Aldrich
Ammonia solution, 2.0 M in isopropanol
Sigma-Aldrich
Formaldehído solution, tested according to Ph. Eur.