Merck
Glioma cells with the IDH1 mutation modulate metabolic fractional flux through pyruvate carboxylase.
PloS one (2014-09-23)
Jose L Izquierdo-Garcia, Larry M Cai, Myriam M Chaumeil, Pia Eriksson, Aaron E Robinson, Russell O Pieper, Joanna J Phillips, Sabrina M Ronen
RESUMEN
Over 70% of low-grade gliomas carry a heterozygous R132H mutation in the gene coding for isocitrate dehydrogenase 1 (IDH1). This confers the enzyme with the novel ability to convert α-ketoglutarate to 2-hydroxyglutarate, ultimately leading to tumorigenesis. The major source of 2-hydroxyglutarate production is glutamine, which, in cancer, is also a source for tricarboxylic acid cycle (TCA) anaplerosis. An alternate source of anaplerosis is pyruvate flux via pyruvate carboxylase (PC), which is a common pathway in normal astrocytes. The goal of this study was to determine whether PC serves as a source of TCA anaplerosis in IDH1 mutant cells wherein glutamine is used for 2-hydroxyglutarate production. Immortalized normal human astrocytes engineered to express heterozygous mutant IDH1 or wild-type IDH1 were investigated. Flux of pyruvate via PC and via pyruvate dehydrogenase (PDH) was determined by using magnetic resonance spectroscopy to probe the labeling of [2-¹³C]glucose-derived ¹³C-labeled glutamate and glutamine. Activity assays, RT-PCR and western blotting were used to probe the expression and activity of relevant enzymes. The Cancer Genome Atlas (TCGA) data was analyzed to assess the expression of enzymes in human glioma samples. Compared to wild-type cells, mutant IDH1 cells significantly increased fractional flux through PC. This was associated with a significant increase in PC activity and expression. Concurrently, PDH activity significantly decreased, likely mediated by significantly increased inhibitory PDH phosphorylation by PDH kinase 3. Consistent with the observation in cells, analysis of TCGA data indicated a significant increase in PC expression in mutant IDH-expressing human glioma samples compared to wild-type IDH. Our findings suggest that changes in PC and PDH may be an important part of cellular adaptation to the IDH1 mutation and may serve as potential therapeutic targets.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Óxido de deuterio, 99.9 atom % D
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Bicarbonato de sodio, ACS reagent, ≥99.7%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Bicarbonato de sodio, powder, BioReagent, Molecular Biology, suitable for cell culture, suitable for insect cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Bicarbonato de sodio, ReagentPlus®, ≥99.5%, powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sodium fluoride, ACS reagent, ≥99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Magnesium chloride, ≥98%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
L-Glutamina, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Óxido de deuterio, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Piruvato sódico, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
L-Glutamina, ReagentPlus®, ≥99% (HPLC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Piruvato sódico, ReagentPlus®, ≥99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Bicarbonato de sodio, anhydrous, free-flowing, Redi-Dri, ACS reagent, ≥99.7%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Bicarbonato de sodio, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E500, 99.0-100.5%, powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
En este momento no podemos mostrarle ni los precios ni la disponibilidad
SAFC
L-Glutamina
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Fosfato de 4-nitrofenilo disodium salt hexahydrate, suitable for enzyme immunoassay, ≥99.0% (enzymatic)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sodium fluoride, ReagentPlus®, ≥99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sustrato de fosfatasa, 5 mg tablets
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Óxido de deuterio, 99.9 atom % D, contains 0.75 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Bicarbonato de sodio, BioXtra, 99.5-100.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Fosfato de 4-nitrofenilo disodium salt hexahydrate, tablet
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Fosfato de 4-nitrofenilo disodium salt hexahydrate, powder, BioReagent, suitable for cell culture, ≥97%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Piruvato sódico, powder, BioXtra, suitable for mouse embryo cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Magnesium chloride, suitable for insect cell culture, BioReagent, ≥97.0%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
USP
Bicarbonato de sodio, United States Pharmacopeia (USP) Reference Standard
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sustrato de fosfatasa, powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
L-Glutamina, BioUltra, ≥99.5% (NT)
En este momento no podemos mostrarle ni los precios ni la disponibilidad