Merck
Molecularly engineered self-assembling membranes for cell-mediated degradation.
Advanced healthcare materials (2014-11-22)
Daniela S Ferreira, Yi-An Lin, Honggang Cui, Jeffrey A Hubbell, Rui L Reis, Helena S Azevedo
RESUMEN
The use of peptide engineering to develop self-assembling membranes that are responsive to cellular enzyme activities is reported. The membranes are obtained by combining hyaluronan (HA) and a rationally designed peptide amphiphile (PA) containing a proteolytic domain (GPQGIWGQ octapeptide) sensitive to matrix metalloproteinase-1 (MMP-1). Insertion of an octapeptide in a typical PA structure does not disturb its self-assembly into fibrillar nanostructures neither the ability to form membranes with HA. In vitro enzymatic degradation with hyaluronidase and MMP-1 shows that membranes containing the MMP-1 substrate exhibit enhanced enzymatic degradation, compared with control membranes (absence of MMP-1 cleavable peptide or containing a MMP-1 insensitive sequence), being completely degraded after 7 days. Cell viability and proliferation is minimally affected by the enzymatically cleavable functionality of the membrane, but the presence of MMP-1 cleavable sequence does stimulate the secretion of MMP-1 by fibroblasts and interfere with matrix deposition, particularly the deposition of collagen. By showing cell-responsiveness to biochemical signals presented on self-assembling membranes, this study highlights the ability of modulating certain cellular activities through matrix engineering. This concept can be further explored to understand the cellular remodeling process and as a strategy to develop artificial matrices with more biomimetic degradation for tissue engineering applications.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido trifluoroacético, ReagentPlus®, 99%
Sigma-Aldrich
Ácido trifluoroacético, suitable for HPLC, ≥99.0%
Sigma-Aldrich
Ácido trifluoroacético, puriss. p.a., suitable for HPLC, ≥99.0% (GC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Formaldehído solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehído solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehído solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Palmitic acid, ≥99%
Sigma-Aldrich
Ácido trifluoroacético, ≥99%, for protein sequencing
Sigma-Aldrich
Piperidine, ReagentPlus®, 99%
Sigma-Aldrich
Triisopropilsilano, 98%
Sigma-Aldrich
Formaldehído solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Piperidine, ≥99.5%, purified by redistillation
Sigma-Aldrich
Palmitic acid, BioXtra, ≥99%
Sigma-Aldrich
Formaldehído solution, meets analytical specification of USP, ≥34.5 wt. %
Sigma-Aldrich
HBTU, ≥98.0% (T)
Sigma-Aldrich
Piperidine, biotech. grade, ≥99.5%
Supelco
Ácido trifluoroacético, analytical standard
Supelco
Palmitic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Formaldehído solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
Palmitic acid, ≥98%, FCC, FG
USP
Palmitic acid, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Formaldehído solution, tested according to Ph. Eur.
Supelco
Palmitic acid, analytical standard
Sigma-Aldrich
Ácido trifluoroacético, ReagentPlus®, 99%
Sigma-Aldrich
Palmitic acid, ≥98% palmitic acid basis (GC)
Sigma-Aldrich
Piperidine solution, suitable for peptide synthesis, 20% in DMF
Palmitic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Palmitic acid, natural, 98%, FG
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone