Merck
  • Genetic Environment of Plasmid Mediated CTX-M-15 Extended Spectrum Beta-Lactamases from Clinical and Food Borne Bacteria in North-Eastern India.
Genetic Environment of Plasmid Mediated CTX-M-15 Extended Spectrum Beta-Lactamases from Clinical and Food Borne Bacteria in North-Eastern India.
PloS one (2015-09-12)
Supriya Upadhyay, Abbas Hussain, Shweta Mishra, Anand Prakash Maurya, Amitabha Bhattacharjee, Santa Ram Joshi
RESUMEN
The study investigated the presence of CTX-M-15 type extended spectrum beta-lactamases (ESBL), compared their genetic arrangements and plasmid types in gram negative isolates of hospital and food origin in north-east India. From September 2013 to April 2014, a total of 252 consecutive, non-duplicate clinical isolates and 88 gram negative food isolates were selected. Phenotypic and molecular characterization of ESBL genes was performed. Presence of integrons and gene cassettes were analyzed by integrase and 59 base-element PCR respectively. The molecular environments surrounding blaCTX-M and plasmid types were investigated by PCR and PCR-based replicon typing respectively. Transformation was carried out to assess plasmid transfer. Southern blotting was conducted to localize the blaCTX-M-15 genes. DNA fingerprinting was performed by ERIC-PCR. Prevalence of ESBL was found to be 40.8% (103/252) in clinical and 31.8% (28/88) in food-borne isolates. Molecular characterization revealed the presence of 56.3% (58/103) and 53.5% (15/28) blaCTX-M-15 in clinical and food isolates respectively. Strains of clinical and food origin were non-clonal. Replicon typing revealed that IncI1 and IncFII plasmid were carrying blaCTX-M-15 in clinical and food isolates and were horizontally transferable. The ISEcp1 element was associated with blaCTX-M-15 in both clinical and food isolates. The simultaneous presence of resistance determinants in non-clonal isolates of two different groups thus suggests that the microbiota of common food products consumed may serve as a reservoir for some of the drug resistance genes prevalent in human pathogens.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Nitroazul de tetrazolio, ≥90.0% (HPLC)
Sigma-Aldrich
Nitroazul de tetrazolio, powder, electrophoresis grade