Merck
  • Apolar Radical Initiated Conjugated Autoxidizable Triene (ApoCAT) Assay: Effects of Oxidant Locations on Antioxidant Capacities and Interactions.
Apolar Radical Initiated Conjugated Autoxidizable Triene (ApoCAT) Assay: Effects of Oxidant Locations on Antioxidant Capacities and Interactions.
Journal of agricultural and food chemistry (2015-08-15)
Atikorn Panya, Waranya Temthawee, Natthaporn Phonsatta, Danai Charoensuk, Pawinee Deetae, Wonnop Visessanguan, Eric A Decker
RESUMEN
Development of an antioxidant assay explaining antioxidant behaviors in complex food systems has been a challenging topic for food scientists. This research aimed to investigate antioxidant capacities and interactions of selected synthetic antioxidants and commercial natural antioxidant extracts using the CAT assay and a newly developed ApoCAT assay, which used water- and lipid-soluble azo radical initiators, respectively. Results suggested that the higher the hydrophobicity of an antioxidant, the higher the antioxidant capacity of an antioxidant observed in the ApoCAT assay. The relationship between the two different assays was explained by the ratio between the ApoCAT and the CAT values. Interestingly, all lipophilic derivatives of the antioxidants exhibited higher ApoCAT/CAT ratios than their hydrophilic derivatives. In the case of the commercial food-grade antioxidants, green tea extract and mixed tocopherols showed a higher antioxidant capacity in the ApoCAT assay than in the CAT assay, while grape seed and rosemary extracts did not show significantly different changes in behaviors in both assays. The study on antioxidant interactions revealed that additive, synergistic, and antagonistic effects between hydrophilic antioxidants and natural extracts, and mixed tocopherols could be observed in both the CAT and the ApoCAT assays, depending on the combined ratios. In most cases, at a particular ratio, the synergistic effect reached the maximum level before suddenly dropping to additive and antagonistic effects in both assays.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Ácido clorhídrico solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ácido L-ascórbico, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Ácido L-ascórbico, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Ácido L-ascórbico, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ácido L-ascórbico, 99%
Sigma-Aldrich
Ácido L-ascórbico, reagent grade, crystalline
Sigma-Aldrich
Ácido clorhídrico, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Ácido gálico, 97.5-102.5% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Supelco
Ácido clorhídrico solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Supelco
2,4,6-Tris(2-pyridyl)-s-triazine, ≥98%
Sigma-Aldrich
Cloruro de hierro (III), anhydrous, powder, ≥99.99% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ácido L-ascórbico, ACS reagent, ≥99%
Sigma-Aldrich
Carbonato de sodio, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Carbonato de sodio, BioXtra, ≥99.0%
Sigma-Aldrich
Ácido clorhídrico solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Ácido L-ascórbico, meets USP testing specifications
Sigma-Aldrich
Cloruro de hierro (III), ≥99.9% trace metals basis
Sigma-Aldrich
Ácido L-ascórbico, reagent grade
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Cloruro de hidrógeno solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Ácido clorhídrico solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Ácido L-ascórbico, puriss. p.a., ACS reagent, reag. ISO, Ph. Eur., 99.7-100.5% (oxidimetric)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ácido L-ascórbico, FCC, FG