Merck
  • Interfacial dilational properties of tea polyphenols and milk proteins with gut epithelia and the role of mucus in nutrient adsorption.
Interfacial dilational properties of tea polyphenols and milk proteins with gut epithelia and the role of mucus in nutrient adsorption.
Food & function (2015-09-04)
Anilda Guri, Yang Li, Milena Corredig
RESUMEN
By interacting with nutrients, the mucus layer covering the intestinal epithelium may mediate absorption. This study aimed to determine possible interactions between epigallocatechin-3-gallate (EGCG), skim milk proteins or their complexes with human intestinal mucin films. The films were extracted from postconfluent monolayers of HT29-MTX, a human intestinal cell line, and a model system was created using drop shape tensiometry. The EGCG uptake tested in vitro on postconfluent Caco-2 cells or co-cultures of Caco-2/HT29-MTX (mucus producing) showed recovery of bioavailable EGCG only for Caco-2 cell monolayers, suggesting an effect of mucus on absorption. Interfacial dilational rheology was employed to characterize the properties of the interface mixed with mucus dispersion. Adsorption of polyphenols greatly enhanced the viscoelastic modulus of the mucus film, showing the presence of interactions between the nutrient molecules and mucus films. On the other hand, in situ digestion of milk proteins using trypsin showed higher surface activities as a result of protein unfolding and competitive adsorption of the hydrolyzed products. There was an increase of viscoelastic modulus over the drop ageing time for the mixed interfaces, indicating the formation of a stiffer interfacial network. These results bring new insights into the role of the mucus layer in nutrient absorption and the interactions of mucus and dairy products.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Cóctel de inhibidores de proteasas, for use with mammalian cell and tissue extracts, DMSO solution
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Metanol, anhydrous, 99.8%
Sigma-Aldrich
L-Glutamina, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Metanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanol, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
L-Glutamina, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Metanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
SAFC
L-Glutamina
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
Metanol, Absolute - Acetone free
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Metanol, BioReagent, ≥99.93%
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
Metanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Tantalum(V) ethoxide, 99.98% trace metals basis
Sigma-Aldrich
L-Glutamina, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
L-Glutamina, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamina
Sigma-Aldrich
Metanol, puriss., meets analytical specification of Ph Eur, ≥99.7% (GC)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
Metanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.
Tantalum(V) ethoxide, packaged for use in deposition systems