Merck
  • Intensive care unit environmental surfaces are contaminated by multidrug-resistant bacteria in biofilms: combined results of conventional culture, pyrosequencing, scanning electron microscopy, and confocal laser microscopy.
Intensive care unit environmental surfaces are contaminated by multidrug-resistant bacteria in biofilms: combined results of conventional culture, pyrosequencing, scanning electron microscopy, and confocal laser microscopy.
The Journal of hospital infection (2015-07-19)
H Hu, K Johani, I B Gosbell, A S W Jacombs, A Almatroudi, G S Whiteley, A K Deva, S Jensen, K Vickery
RESUMEN
Hospital-associated infections cause considerable morbidity and mortality, and are expensive to treat. Organisms causing these infections can be sourced from the inanimate environment around a patient. Could the difficulty in eradicating these organisms from the environment be because they reside in dry surface biofilms? The intensive care unit (ICU) of a tertiary referral hospital was decommissioned and the opportunity to destructively sample clinical surfaces was taken in order to investigate whether multidrug-resistant organisms (MDROs) had survived the decommissioning process and whether they were present in biofilms. The ICU had two 'terminal cleans' with 500 ppm free chlorine solution; items from bedding, surrounds, and furnishings were then sampled with cutting implements. Sections were sonicated in tryptone soya broth and inoculated on to chromogenic plates to demonstrate MDROs, which were confirmed with the Vitek2 system. Genomic DNA was extracted directly from ICU samples, and subjected to polymerase chain reaction (PCR) for femA to detect Staphylococcus aureus and the microbiome by bacterial tag-encoded FLX amplicon pyrosequencing. Confocal laser scanning microscopy (CLSM) and scanning electron microscopy (SEM) were performed on environmental samples. Multidrug-resistant bacteria were cultured from 52% (23/44) of samples cultured. S. aureus PCR was positive in 50%. Biofilm was demonstrated in 93% (41/44) of samples by CLSM and/or SEM. Pyrosequencing demonstrated that the biofilms were polymicrobial and contained species that had multidrug-resistant strains. Dry surface biofilms containing MDROs are found on ICU surfaces despite terminal cleaning with chlorine solution. How these arise and how they might be removed requires further study.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Dodecilsulfatosódico, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Alcohol etílico puro, 200 proof, anhydrous, ≥99.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dodecilsulfatosódico, ≥99.0% (GC), dust-free pellets
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dodecilsulfatosódico, ACS reagent, ≥99.0%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, Molecular Biology, 10% in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Alcohol etílico puro, 190 proof, ACS spectrophotometric grade, 95.0%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dodecilsulfatosódico, ReagentPlus®, ≥98.5% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 25% in H2O, specially purified for use as an electron microscopy fixative
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Glutaraldehyde solution, Grade II, 25% in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Glutaraldehyde solution, 50 wt. % in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dodecilsulfatosódico, BioUltra, Molecular Biology, ≥99.0% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
Dodecilsulfatosódico, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dodecilsulfatosódico, BioXtra, ≥99.0% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dodecilsulfatosódico, ≥98.0% (GC)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 50% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dodecilsulfatosódico, 92.5-100.5% based on total alkyl sulfate content basis
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 70% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dodecilsulfatosódico, ≥90% ((Assay))
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Glutaraldehyde solution, Grade I, 8% in H2O, specially purified for use as an electron microscopy fixative or other sophisticated use
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Glutaric dialdehyde solution, 50 wt. % in H2O, FCC
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dodecilsulfatosódico, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Glutaraldehyde solution, 50% in H2O, suitable for photographic applications
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Dodecilsulfatosódico, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
En este momento no podemos mostrarle ni los precios ni la disponibilidad