Merck
Ubc9 Impairs Activation of the Brown Fat Energy Metabolism Program in Human White Adipocytes.
Molecular endocrinology (Baltimore, Md.) (2015-07-21)
Sean M Hartig, David A Bader, Kathleen V Abadie, Massoud Motamed, Mark P Hamilton, Weiwen Long, Brian York, Michaela Mueller, Martin Wagner, Michael Trauner, Lawrence Chan, Mandeep Bajaj, David D Moore, Michael A Mancini, Sean E McGuire
RESUMEN
Insulin resistance and type 2 diabetes mellitus (T2DM) result from an inability to efficiently store and catabolize surplus energy in adipose tissue. Subcutaneous adipocytes protect against insulin resistance and T2DM by coupling differentiation with the induction of brown fat gene programs for efficient energy metabolism. Mechanisms that disrupt these programs in adipocytes are currently poorly defined, but represent therapeutic targets for the treatment of T2DM. To gain insight into these mechanisms, we performed a high-throughput microscopy screen that identified ubiquitin carrier protein 9 (Ubc9) as a negative regulator of energy storage in human sc adipocytes. Ubc9 depletion enhanced energy storage and induced the brown fat gene program in human sc adipocytes. Induction of adipocyte differentiation resulted in decreased Ubc9 expression commensurate with increased brown fat gene expression. Thiazolidinedione treatment reduced the interaction between Ubc9 and peroxisome proliferator-activated receptor (PPAR)γ, suggesting a mechanism by which Ubc9 represses PPARγ activity. In support of this hypothesis, Ubc9 overexpression remodeled energy metabolism in human sc adipocytes by selectively inhibiting brown adipocyte-specific function. Further, Ubc9 overexpression decreased uncoupling protein 1 expression by disrupting PPARγ binding at a critical uncoupling protein 1 enhancer region. Last, Ubc9 is significantly elevated in sc adipose tissue isolated from mouse models of insulin resistance as well as diabetic and insulin-resistant humans. Taken together, our findings demonstrate a critical role for Ubc9 in the regulation of sc adipocyte energy homeostasis.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Formaldehído solution, Molecular Biology, 36.5-38% in H2O
Sigma-Aldrich
Formaldehído solution, ACS reagent, 37 wt. % in H2O, contains 10-15% Methanol as stabilizer (to prevent polymerization)
SAFC
Formaldehído solution, contains 10-15% methanol as stabilizer, 37 wt. % in H2O
Sigma-Aldrich
Cloruro de amonio, ReagentPlus®, ≥99.5%
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
Sigma-Aldrich
Cloruro de amonio, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Formaldehído solution, Molecular Biology, BioReagent, ≥36.0% in H2O (T)
Sigma-Aldrich
Cloruro de amonio, 99.998% trace metals basis
Sigma-Aldrich
Formaldehído solution, meets analytical specification of USP, ≥34.5 wt. %
Supelco
Formaldehído solution, stabilized with methanol, ~37 wt. % in H2O, certified reference material
Sigma-Aldrich
BIS-TRIS, BioPerformance Certified, suitable for cell culture, suitable for insect cell culture, ≥98.0%
Sigma-Aldrich
BIS-TRIS, BioXtra, ≥98.0% (titration)
Sigma-Aldrich
Cloruro de amonio, 99.99% trace metals basis
Sigma-Aldrich
BIS-TRIS, BioUltra, ≥99.0% (NT)
Sigma-Aldrich
Cloruro de amonio, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, 99.5-100.5% (calc. to the dried substance)
Sigma-Aldrich
Cloruro de amonio, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Formaldehyde-12C solution, 20% in H2O, 99.9 atom % 12C
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)