Merck
Establishment of LC-MS methods for the analysis of palmitoylated surfactant proteins.
Journal of lipid research (2015-05-30)
Takeshi Harayama, Hideo Shindou, Yoshihiro Kita, Eiji Otsubo, Kazushige Ikeda, Shoichi Chida, Timothy E Weaver, Takao Shimizu
RESUMEN
The surfactant proteins (SPs), SP-B and SP-C, are important components of pulmonary surfactant involved in the reduction of alveolar surface tension. Quantification of SP-B and SP-C in surfactant drugs is informative for their quality control and the evaluation of their biological activity. Western blot analysis enabled the quantification of SP-B, but not SP-C, in surfactant drugs. Here, we report a new procedure involving chemical treatments and LC-MS to analyze SP-C peptides. The procedure enabled qualitative analysis of SP-C from different species with discrimination of the palmitoylation status and the artificial modifications that occur during handling and/or storage. In addition, the method can be used to estimate the total amount of SP-C in pulmonary surfactant drugs. The strategy described here might serve as a prototype to establish analytical methods for peptides that are extremely hydrophobic and behave like lipids. The new method provides an easy measurement of SP-C from various biological samples, which will help the characterization of various experimental animal models and the quality control of surfactant drugs, as well as diagnostics of human samples.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Palmitoyl chloride, 98%