Merck
  • Exploring early and late Toxoplasma gondii strain RH infection by two-dimensional immunoblots of chicken immunoglobulin G and M profiles.
Exploring early and late Toxoplasma gondii strain RH infection by two-dimensional immunoblots of chicken immunoglobulin G and M profiles.
PloS one (2015-03-25)
Saeed El-Ashram, Ximeng Sun, Qing Yin, Xianyong Liu, Xun Suo
RESUMEN
Toxoplasma gondii is an intracellular apicomplexan parasite infecting warm-blooded vertebrate hosts, with only early infection stage being contained with drugs. But diagnosis differencing early and late infection was not available. In the present investigation, 2-dimensional immunobloting was used to explore early and late infections in chickens. The protein expression of T. gondii was determined by image analysis of the tachyzoites proteome separated by standard-one and conventional two-dimentional gel polyacrylamide electrophoresis (2D- PAGE). Pooled gels were prepared from tachyzoites of T. gondii. A representative gel spanning a pH range of 3-10 of the tachyzoite proteome consisted of 1306 distinct polypeptide spots. Two-dimensional electrophoresis (2-DE) combined with 2-DE immunoblotting was used to resolve and compare immunoglobulins (Igs) M & G patterns against Toxoplasma gondii strain RH (mouse virulent strain). Total tachyzoite proteins of T. gondii were separated by two-dimensional gel electrophoresis and analyzed by Western blotting for their reactivity with the 7 and 56 days post-infection (dpi) SPF chicken antisera. Different antigenic determinant patterns were detected during analysis with M and G immunoglobulins. Of the total number of polypeptide spots analyzed (1306 differentially expressed protein spots), 6.97% were identified as having shared antigenic polypeptide spots on immunoblot profiles with IgG and IgM antibodies regardless the time after infection. Furthermore, some of the immunoreactive polypeptide spots seemed to be related to the stage of infection. Interestingly, we found natural antibodies to toxoplasmic antigens, in addition to the highly conserved antigenic determinants that reacted with non-specific secondary antibody; goat anti-chicken IgG antibodies conjugated with horseradish peroxidase. In conclusion, unique reactive polypeptide spots are promising candidates for designation of molecular markers to discriminate early and late chicken infection.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Glicina, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glicina, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Metanol, anhydrous, 99.8%
Sigma-Aldrich
Sodium phosphate, 96%
Sigma-Aldrich
Glicina, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glicina, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
SAFC
Glicina
Sigma-Aldrich
Glicina, BioXtra, ≥99% (titration)
Sigma-Aldrich
Glicina, ACS reagent, ≥98.5%
Sigma-Aldrich
Glicina, 99%, FCC
Sigma-Aldrich
Glicina, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Sigma-Aldrich
Glicina, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Metanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol solution, suitable for NMR (reference standard), 4% in methanol-d4 (99.8 atom % D), NMR tube size 3 mm × 8 in.