Merck
  • On the interaction of Helicobacter pylori NikR, a Ni(II)-responsive transcription factor, with the urease operator: in solution and in silico studies.
On the interaction of Helicobacter pylori NikR, a Ni(II)-responsive transcription factor, with the urease operator: in solution and in silico studies.
Journal of biological inorganic chemistry : JBIC : a publication of the Society of Biological Inorganic Chemistry (2015-07-25)
Luca Mazzei, Olena Dobrovolska, Francesco Musiani, Barbara Zambelli, Stefano Ciurli
RESUMEN
Helicobacter pylori (Hp) is a carcinogen that relies on Ni(II) to survive in the extreme pH conditions of the human guts. The regulation of genes coding for Ni(II) enzymes and proteins is effected by the nickel-responsive transcription factor NikR, composed of a DNA-binding domain (DBD) and a metal-binding domain (MBD). The scope of this study is to obtain the molecular details of the HpNikR interaction with the urease operator OP ureA , in solution. The size of the full-length protein prevents the characterization of the HpNikR-OP ureA interaction using NMR. We thus investigated the two separate domains of HpNikR. The conservation of their oligomeric state was established by multiple-angle light scattering. Isothermal calorimetric titrations indicated that the thermodynamics of Ni(II) binding to the isolated MBD is independent of the presence of the adjacent DBDs. The NMR spectra of the isolated DBD support considerable conservation of its structural properties. The spectral perturbations induced on the DBD by OP ureA provided information useful to calculate a structural model of the HpNikR-OP ureA complex using a docking computational protocol. The NMR assignment of the residues involved in the protein-DNA interaction represents a starting point for the development of drugs potentially able to eradicate H. pylori infections. All evidences so far collected, in this and previous studies, consistently indicate that binding of Ni(II) to the MBD increases the HpNikR-DNA affinity by modulating the dynamic, and not the structural, properties of the protein, suggesting that the formation of a stable complex relies upon an induced fit mechanism.
MATERIALES
Número de producto
Marca
Descripción del producto
Sigma-Aldrich
Óxido de deuterio, 99.9 atom % D
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Agua, Nuclease-Free Water, for Molecular Biology
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
HEPES, ≥99.5% (titration)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Agua, sterile-filtered, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
DL-Ditiotreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Cloruro de sodio, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Cloruro de sodio, BioXtra, ≥99.5% (AT)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Cloruro de sodio, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Supelco
DL-Ditiotreitol solution, 1 M in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Óxido de deuterio, 99.9 atom % D, contains 0.05 wt. % 3-(trimethylsilyl)propionic-2,2,3,3-d4 acid, sodium salt
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
BIS-TRIS, ≥98.0% (titration)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Agua, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sodium chloride solution, 5 M
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Agua, Molecular Biology, sterile filtered
En este momento no podemos mostrarle ni los precios ni la disponibilidad
SAFC
Sodium chloride solution, 5 M
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Cloruro de sodio, BioUltra, Molecular Biology, ≥99.5% (AT)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Cloruro de sodio, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Cloruro de sodio, 99.999% trace metals basis
En este momento no podemos mostrarle ni los precios ni la disponibilidad
Sigma-Aldrich
Agua, BioPerformance Certified
En este momento no podemos mostrarle ni los precios ni la disponibilidad